Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-10207REA 876 CHF
BOSSBS-10207R
Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody
Antibodies
Respiratory syncytial virus (RSV) is a major cause of respiratory illness in young children. RSV infection produces a variety of signs and symptoms involving different areas of the respiratory tract, from the nose to the lungs. RSV is a negative sense, enveloped RNA virus. The virion is variable in shape and size with average diameter of between 120 and 300 nm.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RSV Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR