Anti-NPY Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-10071REA 920 CHF
BOSSBS-10071R
Anti-NPY Rabbit Polyclonal Antibody
Antibodies
The onset of angiogenesis is believed to be an early event in tumorigenesis and may facilitate tumor progression and metastasis. Several growth factors with angiogenic activity have been described and include fibroblast growth factor (FGF), platelet derived growth factor (PDGF) and vascular endothelial growth factors (VEGFs). The VEGF protein family is comprised of VEGF, VEGF-B, VEGF-C and VEGF-D, all of which may exhibit angiogenic function in vivo. VEGF-B, which exists as two alternatively spliced isoforms known as VEGF-B167 and VEGF-B186, is abundantly expressed in heart and skeletal muscle and is frequently co-expressed with VEGF. VEGF-C binds to and specifically activates Flt-4 and Flk-1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: NPY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR