Anti-CIDEB Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-0716REA 920 CHF
BOSSBS-0716R
Anti-CIDEB Rabbit Polyclonal Antibody
Antibodies
Activates apoptosis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CIDEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR