Anti-c-phycocyanin Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-0697REA 894 CHF
BOSSBS-0697R
Anti-c-phycocyanin Rabbit Polyclonal Antibody
Antibodies
C-Phycocyanin (CPC), a pigment-protein complex from the light-harvesting phycobiliprotein family.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: c-phycocyanin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR