Anti-Dmt1 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-0653REA 920 CHF
BOSSBS-0653R
Anti-Dmt1 Rabbit Polyclonal Antibody
Antibodies
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Dmt1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR