Anti-KAT2A Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9713EA 730 CHF
BSBTPB9713
Anti-KAT2A Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Histone acetyltransferase KAT2A(KAT2A) detection. Tested with WB in Human.

Type: Primary
Antigen: KAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR