Anti-F2RL1 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9700EA 730 CHF
BSBTPB9700
Anti-F2RL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Proteinase-activated receptor 2(F2RL1) detection. Tested with WB in Human.

Type: Primary
Antigen: F2RL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR