Anti-B3GNT8 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9686EA 693 CHF
BSBTPB9686
Anti-B3GNT8 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8(B3GNT8) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: B3GNT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR