Anti-CDC6 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29498EA 622 CHF
ABNOPAB29498
Anti-CDC6 Rabbit Polyclonal Antibody
Antibodies
CDC6 polyclonal antibody raised against recombinant human CDC6.

Type: Primary
Antigen: CDC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR