Anti-FAS Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29420EA 622 CHF
ABNOPAB29420
Anti-FAS Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant human FAS.

Type: Primary
Antigen: FAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR