Anti-SELP Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29415EA 622 CHF
ABNOPAB29415
Anti-SELP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant human SELP.

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR