Anti-AHSA1 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29351EA 622 CHF
ABNOPAB29351
Anti-AHSA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant human AHSA1.

Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR