Anti-HSPA9 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB29350EA 622 CHF
ABNOPAB29350
Anti-HSPA9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant human HSPA9.

Type: Primary
Antigen: HSPA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR