Anti-Virus HA Mouse Monoclonal Antibody [clone: 4B9]

Supplier: Abnova
ABNOMAB6542-M01EA 488 CHF
ABNOMAB6542-M01
Anti-Virus HA Mouse Monoclonal Antibody [clone: 4B9]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HA (Influenza A virus H5N1).

Type: Primary
Antigen: Virus HA
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR