Anti-Virus HA Mouse Monoclonal Antibody [clone: 1A9]

Supplier: Abnova
ABNOMAB4953-M10EA 488 CHF
ABNOMAB4953-M10
Anti-Virus HA Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Mouse monoclonal antibody raised against partial recombinant HA (Influenza A virus H7N1).

Type: Primary
Antigen: Virus HA
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR