Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 2F6]

Supplier: Abnova
ABNOH00023495-M06EA 473 CHF
ABNOH00023495-M06
Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 2F6]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFRSF13B.

Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR