Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 8D4]

Supplier: Abnova
ABNOH00027242-M63EA 473 CHF
ABNOH00027242-M63
Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 8D4]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFRSF21.

Type: Primary
Antigen: TNFRSF21
Clonality: Monoclonal
Clone: 8D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR