Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 4C9]

Supplier: Abnova
ABNOH00008764-M15EA 473 CHF
ABNOH00008764-M15
Anti-TNFRSF14 Mouse Monoclonal Antibody [clone: 4C9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFRSF14.

Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR