Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 4F1]

Supplier: Abnova
ABNOH00008792-M01EA 473 CHF
ABNOH00008792-M01
Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 4F1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFRSF11A.

Type: Primary
Antigen: TNFRSF11A
Clonality: Monoclonal
Clone: 4F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR