Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 2H3]

Supplier: Abnova
ABNOH00009966-M06EA 473 CHF
ABNOH00009966-M06
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 2H3]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFSF15.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR