Anti-EDA Mouse Monoclonal Antibody [clone: 4D9]

Supplier: Abnova
ABNOH00001896-M16EA 473 CHF
ABNOH00001896-M16
Anti-EDA Mouse Monoclonal Antibody [clone: 4D9]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant EDA.

Type: Primary
Antigen: EDA
Clonality: Monoclonal
Clone: 4D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR