Anti-CD86 Mouse Monoclonal Antibody [clone: 4H2]

Supplier: Abnova
ABNOH00000942-M05EA 473 CHF
ABNOH00000942-M05
Anti-CD86 Mouse Monoclonal Antibody [clone: 4H2]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant CD86.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR