Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 7A6]

Supplier: Abnova
ABNOMAB1180-M04EA 488 CHF
ABNOMAB1180-M04
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 7A6]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10A.

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 7A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR