Anti-CD276 Mouse Monoclonal Antibody [clone: 2E12]

Supplier: Abnova
ABNOH00080381-M15EA 473 CHF
ABNOH00080381-M15
Anti-CD276 Mouse Monoclonal Antibody [clone: 2E12]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant CD276.

Type: Primary
Antigen: CD276
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR