Anti-LTA Mouse Monoclonal Antibody [clone: 3C1]

Supplier: Abnova
ABNOH00004049-M48EA 473 CHF
ABNOH00004049-M48
Anti-LTA Mouse Monoclonal Antibody [clone: 3C1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant LTA.

Type: Primary
Antigen: LTA
Clonality: Monoclonal
Clone: 3C1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR