Anti-CD274 Mouse Monoclonal Antibody [clone: 3F4]

Supplier: Abnova
ABNOH00029126-M01EA 473 CHF
ABNOH00029126-M01
Anti-CD274 Mouse Monoclonal Antibody [clone: 3F4]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant CD274.

Type: Primary
Antigen: CD274
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR