Anti-CCDC69 Mouse Monoclonal Antibody [clone: 4G6]

Supplier: Abnova
ABNOH00026112-M05EA 473 CHF
ABNOH00026112-M05
Anti-CCDC69 Mouse Monoclonal Antibody [clone: 4G6]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant CCDC69.

Type: Primary
Antigen: CCDC69
Clonality: Monoclonal
Clone: 4G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR