Anti-LY6G Rat Monoclonal Antibody [clone: RB6-8C5] (PE (Phycoerythrin)/Cy7®)

Supplier: Abnova
ABNOMAB12236EA 503 CHF
ABNOMAB12236
Anti-LY6G Rat Monoclonal Antibody [clone: RB6-8C5] (PE (Phycoerythrin)/Cy7®)
Antibodies
Rat monoclonal antibody raised against Ly6g.

Type: Secondary
Antigen: bone marrow cells
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PE (Phycoerythrin)/Cy7
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR