Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL1] (FITC (Fluorescein Isothiocyanate))

Supplier: Abnova
ABNOMAB12231EA 479 CHF
ABNOMAB12231
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL1] (FITC (Fluorescein Isothiocyanate))
Antibodies
Mouse monoclonal antibody raised against CD45RO.

Type: Secondary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR