Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (PE (Phycoerythrin)/Cy7®)

Supplier: Abnova
ABNOMAB12210EA 503 CHF
ABNOMAB12210
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (PE (Phycoerythrin)/Cy7®)
Antibodies
Rat monoclonal antibody raised against Cd4.

Type: Secondary
Antigen: Cd4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: PE (Phycoerythrin)/Cy7
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR