Anti-TMEFF2 Mouse Polyclonal Antibody

Supplier: Abnova
ABNOH00023671-B01PEA 473 CHF
ABNOH00023671-B01P
Anti-TMEFF2 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human TMEFF2 protein.

Type: Primary
Antigen: TMEFF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR