Anti-CD62P P-selectin Chicken Polyclonal Antibody (Fluorescein)

Supplier: Agrisera
AGRIIMS09-143-338EA 490 CHF
AGRIIMS09-143-338
Anti-CD62P P-selectin Chicken Polyclonal Antibody (Fluorescein)
Antibodies
Platelets or thrombocytes are the cell fragments circulating in the blood that are involved in the cellular mechanisms of primary hemostasis leading to the formation of blood clots. Dysfunction or low levels of platelets predisposes to bleeding, while high levels, although usually asymptomatic, may increase the risk of thrombosis. Platelets are produced in the bone marrow; the progenitor cell for platelets is the megakaryocyte. It is about twelve times larger than an erythrocyte, possesses a lobed nucleus and sheds platelets into the circulation. Alternative protein names include: CD62P; GMP140; GMRP; Granule membrane protein 140; Granulocyte membrane protein; GRMP; LECAM 3; LECAM3; Leukocyte endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule external membrane protein; PSEL; SELP,Platelets or thrombocytes are the cell fragments circulating in the blood that are involved in the cellular mechanisms of primary hemostasis leading to the formation of blood clots. Dysfunction or low levels of platelets predisposes to bleeding, while high levels, although usually asymptomatic, may increase the risk of thrombosis. Platelets are produced in the bone marrow; the progenitor cell for platelets is the megakaryocyte. It is about twelve times larger than an erythrocyte, possesses a lobed nucleus and sheds platelets into the circulation. Alternative protein names include: CD62P; GMP140; GMRP; Granule membrane protein 140; Granulocyte membrane protein; GRMP; LECAM 3; LECAM3; Leukocyte endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule external membrane protein; PSEL; SELP

Type: Primary
Antigen: CD62P
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Chicken
Isotype:
Reactivity: A. thaliana, H. vulgare, Populus sp., T. aestivum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR