Anti-TKL1 Rabbit Polyclonal Antibody

Supplier: Agrisera
AGRIAS152903EA 466 CHF
AGRIAS152903
Anti-TKL1 Rabbit Polyclonal Antibody
Antibodies
TKL-1 (Transketolase-1, chloroplastic) is involved in the pathway of Calvin cycle. This enzyme is catalyzing the reversible transfer of a two-carbon ketol group from fructose-6-phopshate or sedoheptuloze-7-phosphate to glyceraldehyde-3-phosphate to yield xylulose-5-phosphate and erythrose-4-phosphate or ribose-5-phosphate. AtTKL1 is expressed in highest levels in photosynthetic tissue, while AtTKL2 is expressed mainly during embryo development. Alternative name: TK.,TKL-1 (Transketolase-1, chloroplastic) is involved in the pathway of Calvin cycle. This enzyme is catalyzing the reversible transfer of a two-carbon ketol group from fructose-6-phopshate or sedoheptuloze-7-phosphate to glyceraldehyde-3-phosphate to yield xylulose-5-phosphate and erythrose-4-phosphate or ribose-5-phosphate. AtTKL1 is expressed in highest levels in photosynthetic tissue, while AtTKL2 is expressed mainly during embryo development. Alternative name: TK.

Type: Primary
Antigen: TKL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Pisum sativum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR