Anti-PRN2 Rabbit Polyclonal Antibody

Supplier: Agrisera
AGRIAS142794EA 466 CHF
AGRIAS142794
Anti-PRN2 Rabbit Polyclonal Antibody
Antibodies
PRN2 (PIRIN) belongs to a functionally diverse cupin protein superfamily with four family members of Arabidopsis thaliana PRN proteins. ,PRN2 (PIRIN) belongs to a functionally diverse cupin protein superfamily with four family members of Arabidopsis thaliana PRN proteins.

Type: Primary
Antigen: PRN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR