Anti-IRF2 Rabbit Polyclonal Antibody
Supplier: Boster
BSBTPB9645EA 730 CHF
BSBTPB9645
Anti-IRF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Interferon regulatory factor 2(IRF2) detection. Tested with WB in Human;Rat.
IRF2 (interferon regulatory factor 2) is a member of the interferon regulatory transcription factor (IRF) family. The IRF2 gene is mapped on 4q35.1. When the IRF2 gene was overexpressed in NIH 3T3 cells, the cells became transformed and displayed enhanced tumorigenicity in nude mice. One IRF binding site was found within the IRF2 promoter, and expression of the IRF2 gene was affected by both transient and stable IRF1 expression. IRF2 competitively inhibits the IRF1-mediated transcriptional activation of interferons alpha and beta, and presumably other genes that employ IRF1 for transcription activation. However, IRF2 also functions as a transcriptional activator of histone H4. Irf2 was required to prevent NK-cell apoptosis and keep immature NK cells alive, thus promoting NK-cell maturation and their supply to peripheral blood.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.