Anti-SOX4 Antibody

Supplier: Boster
BSBTPB9618EA 730 CHF
BSBTPB9618
Anti-SOX4 Antibody
Antibodies
Rabbit IgG polyclonal antibody for Transcription factor SOX-4(SOX4) detection. Tested with WB in Human.

Type: Primary
Antigen: SOX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR