Anti-HNF1 beta Antibody

Supplier: Boster
BSBTPB9597EA 730 CHF
BSBTPB9597
Anti-HNF1 beta Antibody
Antibodies
Rabbit IgG polyclonal antibody for Hepatocyte nuclear factor 1-beta(HNF1B) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: HNF1 beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR