Anti-FUT1 Antibody

Supplier: Boster
BSBTPB9593EA 693 CHF
BSBTPB9593
Anti-FUT1 Antibody
Antibodies
Rabbit IgG polyclonal antibody for Galactoside 2-alpha-L-fucosyltransferase 1(FUT1) detection. Tested with WB, IHC-P in Human;Mouse; Rat.

Type: Primary
Antigen: FUT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR