Anti-Stathmin 1 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9560EA 693 CHF
BSBTPB9560
Anti-Stathmin 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Stathmin(STMN1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Stathmin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR