Anti-IL6 Mouse Monoclonal Antibody Pair
Supplier: Abnova
ABNOH00003569-AP41EA 1010 CHF
ABNOH00003569-AP41
Anti-IL6 Mouse Monoclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human IL6.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-IL6, IgG2a kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-IL6, IgG2b kappa (50 ?g);