Anti-USP9X Mouse Monoclonal Antibody (Biotin)

Supplier: Abnova
ABNOH00008239-AP41EA 1010 CHF
ABNOH00008239-AP41
Anti-USP9X Mouse Monoclonal Antibody (Biotin)
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human USP9X.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-USP9X, IgG1 kappa (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-USP9X, IgG1 kappa (50 ?g);

Type:
Antigen: USP9X
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR