Anti-PPM1B Mouse Monoclonal Antibody

Supplier: Abnova
ABNOH00005495-PW1EA 1010 CHF
ABNOH00005495-PW1
Anti-PPM1B Mouse Monoclonal Antibody
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-PPM1B (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-PPM1B (50 ?g)

Type:
Antigen: PPM1B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR