Anti-HSPA13 Polyclonal Antibody Pair

Supplier: Abnova
ABNOH00006782-AP22EA 1010 CHF
ABNOH00006782-AP22
Anti-HSPA13 Polyclonal Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human HSPA13.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-HSPA13 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-HSPA13 (20 µg);

Type:
Antigen: HSPA13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR