Anti-S100 B Mouse Monoclonal Antibody (Biotin)

Supplier: Abnova
ABNOH00006285-AP43EA 1010 CHF
ABNOH00006285-AP43
Anti-S100 B Mouse Monoclonal Antibody (Biotin)
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human S100B.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-S100B (100 ?g); 2. Detection Antibody: biotinylated Mouse Monoclonal Anti-S100B, IgG1 kappa (50 ?g);

Type:
Antigen: S100 B
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR