Anti-NDUFS3 Polyclonal Antibody Pair
Supplier: Abnova
ABNOH00004722-PW1EA 1010 CHF
ABNOH00004722-PW1
Anti-NDUFS3 Polyclonal Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-NDUFS3 (300 µl); 2. Antibody pair for Western Blot: Mouse Polyclonal Anti-NDUFS3 (50 µl)
Type:
Antigen: NDUFS3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human