Anti-IFITM3 Antibody Pair

Supplier: Abnova
ABNOH00010410-AP11EA 1010 CHF
ABNOH00010410-AP11
Anti-IFITM3 Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human IFITM3.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-IFITM3 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-IFITM3, IgG1 kappa (20 µg);

Type:
Antigen: IFITM3
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR