Anti-HOXB9 Antibody Pair

Supplier: Abnova
ABNOH00003219-AP11EA 1010 CHF
ABNOH00003219-AP11
Anti-HOXB9 Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human HOXB9.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-HOXB9 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-HOXB9, IgG2b kappa (20 µg);

Type:
Antigen: HOXB9
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR