Anti-PGR + PGR Antibody Pair

Supplier: Abnova
ABNODP0290EA 882 CHF
ABNODP0290
Anti-PGR + PGR Antibody Pair
Antibodies
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the PGR protein, and the other against the specific S190 phosphorylated site of PGR for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-PGR S190 Rabbit Polyclonal Antibody (20 µl) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PGR Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: PGR + PGR
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR