Anti-CRKL + EGFR Polyclonal Antibody Pair

Supplier: Abnova
ABNODI0003EA 802 CHF
ABNODI0003
Anti-CRKL + EGFR Polyclonal Antibody Pair
Antibodies
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CRKL protein, and the other against the EGFR protein for use in Proximity Ligation Assay

Antibody pair set content: 1. CRKL Rabbit purified Polyclonal Antibody (20 µg) 2. EGFR Mouse purified Polyclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CRKL + EGFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR