Anti-PIP5K1C Antibody Pair

Supplier: Abnova
ABNOH00023396-AP51EA 1010 CHF
ABNOH00023396-AP51
Anti-PIP5K1C Antibody Pair
Antibodies
This Set comes with matched Antibody Pair to detect and quantify protein level of human PIP5K1C.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-PIP5K1C (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-PIP5K1C (50 µg);

Type:
Antigen: PIP5K1C
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR